Tetramethylammonium hydroxide, 25 wt% in methanol
Tetramethylammonium hydroxide, 25 wt% in methanol
Tetramethylammonium hydroxide, 25 wt% in methanol
Tetramethylammonium hydroxide, 25 wt% in methanol
Thermo Scientific Chemicals
Tetramethylammonium hydroxide, 25 wt% in methanol
CAS: 75-59-2 | C4H13NO | 91.154 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA138355000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS75-59-2, 67-56-1
Specifications
Chemical Name or MaterialTetramethylammonium hydroxide
Name Note25 wt. % in Methanol
CAS75-59-2
Boiling Point65.0°C
Health Hazard 1Danger
View more
Phase transfer catalyst
RUO – Research Use Only