1,2,3,4-Tetrahydroisoquinoline, 95%
1,2,3,4-Tetrahydroisoquinoline, 95%
1,2,3,4-Tetrahydroisoquinoline, 95%
1,2,3,4-Tetrahydroisoquinoline, 95%
Thermo Scientific Chemicals
1,2,3,4-Tetrahydroisoquinoline, 95%
CAS: 91-21-4 | C9H11N | 133.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA138200250
Price (MYR)
480.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
480.00
BOT
Add to cart
Specifications
Chemical Name or Material1,2,3,4-Tetrahydroisoquinoline
CAS91-21-4
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0