DL-Tartaric acid, 99.5%
DL-Tartaric acid, 99.5%
DL-Tartaric acid, 99.5%
DL-Tartaric acid, 99.5%
Thermo Scientific Chemicals
DL-Tartaric acid, 99.5%
CAS: 133-37-9 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA137875000
Price (MYR)
651.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
651.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDL-Tartaric acid
CAS133-37-9
Melting Point200.0°C to 206.0°C
Health Hazard 1Danger