Check Order Status
2-Amino-6-chlorobenzothiazole, 99%, Thermo Scientific™
Element Search
Structure Search
Thermo Scientific Chemicals
2-Amino-6-chlorobenzothiazole, 99%, Thermo Scientific™
Have Questions?
Change view
Quantity:
10 g
Packaging:
Glass bottle
Catalog number
FSA136400100
Price (MYR)
327.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
327.00
BOT
Add to cart
Specifications
Chemical Name or Material
2-Amino-6-chlorobenzothiazole
CAS
95-24-9
CAS Min %
98.5
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0