View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialActivated charcoal,NORIT* SA 2
Name NoteNORIT™ SA 2, Decolorizing
CAS7440-44-0
Melting Point3652.0°C
ColorBlack