5-Sulfosalicylic Acid Dihydrate, 98%
5-Sulfosalicylic Acid Dihydrate, 98%
5-Sulfosalicylic Acid Dihydrate, 98%
5-Sulfosalicylic Acid Dihydrate, 98%
Thermo Scientific Chemicals
5-Sulfosalicylic Acid Dihydrate, 98%
CAS: 5965-83-3 | C9H14O6S | 250.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA132862500
Price (MYR)
324.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
324.00
BOT
Add to cart
Chemical Identifiers
CAS5965-83-3, 97-05-2
Specifications
Chemical Name or Material5-Sulfosalicylic acid dihydrate
CAS5965-83-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Indicator
RUO – Research Use Only