Sulfanilamide 98%
Sulfanilamide 98%
Sulfanilamide 98%
Thermo Scientific Chemicals
Sulfanilamide 98%
Sulfanilamide, CAS # 63-74-1, is an organic sulfur compound that shows antibacterial activities.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Catalog number FSA132855000
Price (MYR)
901.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
901.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSulfanilamide
TypeSulfanilamide
Name Note98%
Physical FormCrystalline Powder
CAS63-74-1
View more
General Description
  • Sulfanilamide is an organic sulfur compound with antibacterial properties that are structurally similar to the p-aminobenzoic acid (PABA)
  • It also has a competitive inhibitory capacity against dihydropteroate synthetases blocking the synthesis of folic acid
  • Sulfanilamide competes with PABA for the bacterial enzyme dihydropteroate synthase preventing, the incorporation of PABA into dihydrofolic acid, an immediate precursor of folic acid. This culminates in the inhibition of the bacterial folic acid synthesis and also the de novo synthesis of purines and pyrimidines, resulting in cell growth arrest and cell death.
Applications
  • This compound shows antibacterial activity against different species, including gram-positive, gram-negative, and Chlamydia
  • In chemical reactions, this compound can act as an intermediate for the preparation of 2,6-disubstituted anilines by electrophilic substitution
  • Sulfanilamide shows inhibitory activity against the enzyme, carbonic anhydrase.
RUO – Research Use Only