Succinyl Chloride, ca. 95%
Succinyl Chloride, ca. 95%
Succinyl Chloride, ca. 95%
Succinyl Chloride, ca. 95%
Thermo Scientific Chemicals
Succinyl Chloride, ca. 95%
CAS: 543-20-4 | C4H4Cl2O2 | 154.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA132840250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSuccinyl chloride
CAS543-20-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with