Spermine, 97%
Spermine, 97%
Spermine, 97%
Thermo Scientific Chemicals
Spermine, 97%
CAS: 71-44-3 | C10H26N4 | 202.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA132750050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSpermine
Name Note97%
CAS71-44-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement