Sebacic Acid, 98%
Sebacic Acid, 98%
Sebacic Acid, 98%
Thermo Scientific Chemicals
Sebacic Acid, 98%
CAS: 111-20-6 | C10H18O4 | 202.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number FSA132631000
Price (MYR)
198.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
198.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSebacic acid
CAS111-20-6
Melting Point131.0°C to 134.0°C
ColorWhite
EINECS Number203-845-5