1,4-Dihydroxyanthraquinone, 96%
1,4-Dihydroxyanthraquinone, 96%
1,4-Dihydroxyanthraquinone, 96%
1,4-Dihydroxyanthraquinone, 96%
1,4-Dihydroxyanthraquinone, 96%
CAS: 81-64-1 | C14H8O4 | 240.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA132181000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 4-Dihydroxyanthraquinone
Name Note96%
CAS81-64-1
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning