2-Pyrrolidinone, 99%
2-Pyrrolidinone, 99%
2-Pyrrolidinone, 99%
2-Pyrrolidinone, 99%
Thermo Scientific Chemicals
2-Pyrrolidinone, 99%
CAS: 616-45-5 | C4H7NO | 85.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
250 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA132115000
Price (MYR)
387.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
387.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Pyrrolidinone
Name Note98%
CAS616-45-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning