Propargyl bromide, 80 wt.% solution in toluene, stabilized
Propargyl bromide, 80 wt.% solution in toluene, stabilized
Propargyl bromide, 80 wt.% solution in toluene, stabilized
Propargyl bromide, 80 wt.% solution in toluene, stabilized
Thermo Scientific Chemicals
Propargyl bromide, 80 wt.% solution in toluene, stabilized
CAS: 106-96-7 | C3H3Br | 118.961 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
50 mL
Packaging:
Glass bottle
Catalog number FSA131482500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106-96-7, 108-88-3
Specifications
Chemical Name or MaterialPropargyl bromide
ColorWhite
CAS106-96-7
Boiling Point88.0°C to 90.0°C
Health Hazard 1Danger
View more
Alkylation reagent
RUO – Research Use Only