2-Propanethiol, 98%
2-Propanethiol, 98%
2-Propanethiol, 98%
Thermo Scientific Chemicals
2-Propanethiol, 98%
CAS: 75-33-2 | C3H8S | 76.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA131445000
Price (MYR)
572.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
572.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Propanethiol
Name Note98%
CAS75-33-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger