Piperazine, 99%, extra pure
Piperazine, 99%, extra pure
Piperazine, 99%, extra pure
Piperazine, 99%, extra pure
Thermo Scientific Chemicals
Piperazine, 99%, extra pure
CAS: 110-85-0 | C4H10N2 | 86.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
10 kg
Catalog number FSA131295000
Price (MYR)
443.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
443.00
EA
Add to cart
Specifications
Chemical Name or MaterialPiperazine
Name Noteanhydrous, 99%
CAS110-85-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility.
Suspected of damaging