Picolinic acid, 99%
Picolinic acid, 99%
Picolinic acid, 99%
Thermo Scientific Chemicals
Picolinic acid, 99%
CAS: 98-98-6 | C6H5NO2 | 123.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number FSA131171000
Price (MYR)
330.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
330.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPicolinic acid
Name Note99%
CAS98-98-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement