Phenyl sulfide, 99%
Phenyl sulfide, 99%
Phenyl sulfide, 99%
Thermo Scientific Chemicals
Phenyl sulfide, 99%
CAS: 139-66-2 | C12H10S | 186.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA130951000100 mLGlass bottle
Catalog number FSA130951000
Price (MYR)
784.00
BOT
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
784.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenyl sulfide
Name Note99%
CAS139-66-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: