2-Phenylphenol, 99+%
2-Phenylphenol, 99+%
2-Phenylphenol, 99+%
2-Phenylphenol, 99+%
Thermo Scientific Chemicals
2-Phenylphenol, 99+%
CAS: 90-43-7 | C12H10O | 170.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA130765000
Price (MYR)
451.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
451.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Phenylphenol
Name Note99+%
CAS90-43-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning