p-Phenylenediamine, 99+%
p-Phenylenediamine, 99+%
p-Phenylenediamine, 99+%
p-Phenylenediamine, 99+%
Thermo Scientific Chemicals
p-Phenylenediamine, 99+%
CAS: 106-50-3 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Packaging:
Glass bottle
Catalog number FSA130575000
Price (MYR)
646.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
646.00
BOT
Add to cart
Specifications
Chemical Name or Materialp-Phenylenediamine
Name Note99+%
CAS106-50-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
May cause an allergic skin reaction.
Toxic if inhaled.