o-Phenylenediamine, 98%
o-Phenylenediamine, 98%
o-Phenylenediamine, 98%
o-Phenylenediamine, 98%
Thermo Scientific Chemicals
o-Phenylenediamine, 98%
CAS: 95-54-5 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA130552500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialo-Phenylenediamine
CAS95-54-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0