Phenoxazine, 97%
Phenoxazine, 97%
Phenoxazine, 97%
Thermo Scientific Chemicals
Phenoxazine, 97%
CAS: 135-67-1 | C12H9NO | 183.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA130220050
Price (MYR)
1,020.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,020.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenoxazine
CAS135-67-1
Melting Point154.0°C to 159.0°C
ColorGray to Gray-Green
EINECS Number205-210-8