Perylene, 99+%
Perylene, 99+%
Perylene, 99+%
Thermo Scientific Chemicals
Perylene, 99+%
CAS: 198-55-0 | C20H12 | 252.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA130070050
Price (MYR)
911.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
911.00
EA
Add to cart
Specifications
Chemical Name or MaterialPerylene
CAS198-55-0
Melting Point277.0°C to 280.0°C
ColorBrown to Ochre
EINECS Number205-900-9