n-Pentadecane, 99%
n-Pentadecane, 99%
n-Pentadecane, 99%
Thermo Scientific Chemicals
n-Pentadecane, 99%
CAS: 629-62-9 | C15H32 | 212.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA129822500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialn-Pentadecane
Name Note99%
CAS629-62-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger