Palmitic acid, 98%
Palmitic acid, 98%
Palmitic acid, 98%
Thermo Scientific Chemicals
Palmitic acid, 98%
CAS: 57-10-3 | C16H32O2 | 256.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA129702500
Price (MYR)
238.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
238.00
EA
Add to cart
Specifications
Chemical Name or MaterialPalmitic acid
Acidity215 to 233mg KOH/g
CAS57-10-3
Health Hazard 1Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Fatal if swallowed.