Octanoic acid, 99%
Octanoic acid, 99%
Octanoic acid, 99%
Thermo Scientific Chemicals
Octanoic acid, 99%
Octanoic acid, 99%, C8H16O2, CAS Number-124-07-2, 1-heptanecarboxylic acid, caprylic acid, enantic acid, n-caprylic acid, neo-fat 8, n-octanoic acid, n-octoic acid, n-octylic acid, octoic acid, octylic acid, 1L, 385 to 393mg KOH/g, 98.5% min. (GC), 02, 347, 238.0 deg.C, 100.0, CHEBI:28837 | CAS: 124-07-2 | C8H16O2 | 144.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal drum
Catalog number FSA129390251
Price (MYR)
3,407.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Price (MYR)
3,407.00
EA
Add to cart
Specifications
Chemical Name or MaterialOctanoic acid
Acidity385 to 393mg KOH/g
CAS124-07-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger