1,7-Octadiene, 98.5%
1,7-Octadiene, 98.5%
1,7-Octadiene, 98.5%
1,7-Octadiene, 98.5%
Thermo Scientific Chemicals
1,7-Octadiene, 98.5%
CAS: 3710-30-3 | C8H14 | 110.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Catalog number FSA129351000
Price (MYR)
562.00
BOT
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
562.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 7-Octadiene
CAS3710-30-3
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0