Octadecylamine, 90%, tech.
Octadecylamine, 90%, tech.
Octadecylamine, 90%, tech.
Octadecylamine, 90%, tech.
Thermo Scientific Chemicals
Octadecylamine, 90%, tech.
CAS: 124-30-1 | C18H39N | 269.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Catalog number FSA129325000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialOctadecylamine
CAS124-30-1
CAS Min %88.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0