4-Nitrocatechol, 97%
4-Nitrocatechol, 97%
4-Nitrocatechol, 97%
4-Nitrocatechol, 97%
Thermo Scientific Chemicals
4-Nitrocatechol, 97%
CAS: 3316-09-4 | C6H5NO4 | 155.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Packaging:
Glass bottle
Catalog number FSA128600500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitrocatechol
CAS3316-09-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0