4-Nitrobenzonitrile, 97%
4-Nitrobenzonitrile, 97%
4-Nitrobenzonitrile, 97%
4-Nitrobenzonitrile, 97%
Thermo Scientific Chemicals
4-Nitrobenzonitrile, 97%
CAS: 619-72-7 | C7H4N2O2 | 148.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA128481000
Price (MYR)
1,792.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,792.00
EA
Add to cart
Specifications
Chemical Name or Material4-Nitrobenzonitrile
Name Note97%
CAS619-72-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger