p-Nitroacetophenone, 97%
p-Nitroacetophenone, 97%
p-Nitroacetophenone, 97%
p-Nitroacetophenone, 97%
Thermo Scientific Chemicals
p-Nitroacetophenone, 97%
CAS: 100-19-6 | C8H7NO3 | 165.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA128341000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialp-Nitroacetophenone
Name Note97%
CAS100-19-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: