1-Naphthoic acid, 98%
1-Naphthoic acid, 98%
1-Naphthoic acid, 98%
1-Naphthoic acid, 98%
Thermo Scientific Chemicals
1-Naphthoic acid, 98%
CAS: 86-55-5 | C11H8O2 | 172.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA128181000
Price (MYR)
751.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
751.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Naphthoic acid
Name Note98%
CAS86-55-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning