alpha-Naphthoflavone, 97%
alpha-Naphthoflavone, 97%
alpha-Naphthoflavone, 97%
alpha-Naphthoflavone, 97%
Thermo Scientific Chemicals
alpha-Naphthoflavone, 97%
CAS: 604-59-1 | C19H12O2 | 272.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA128160050
Price (MYR)
927.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
927.00
BOT
Add to cart
Specifications
Chemical Name or Materialα-Naphthoflavone
Name Note99.5+%
CAS604-59-1
CAS Min %99.5
CAS Max %100.0
View more
Indicator
RUO – Research Use Only