Starfish
Starfish
Actual product may vary
Thermo Scientific Chemicals
Starfish
Pisaster sp.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)Quantity
FSA1281002506 to 8 in.50 Pk.
ACR1281002506 to 8 in.50 Pk.
Catalog number FSA128100250
Price (MYR)
738.00
BOT
Add to cart
Length (English):
6 to 8 in.
Quantity:
50 Pk.
Price (MYR)
738.00
BOT
Add to cart
  • Easy-to-dissect Asteroidea-starfish from the genus Pisaster is a representative invertebrate deuterosome
  • Single-injected specimens have latex in the water vascular system
  • Select vacuum packed or in pails
Specifications
TypeStarfish
Injection TypePlain
Length (English)6 to 8 in.
Length (Metric)15.2 to 20.3 cm
Quantity50 Pk.
PackagingPail
Unit SizeBOT