2-Naphthaldehyde, 98%
2-Naphthaldehyde, 98%
2-Naphthaldehyde, 98%
2-Naphthaldehyde, 98%
Thermo Scientific Chemicals
2-Naphthaldehyde, 98%
CAS: 66-99-9 | C11H8O | 156.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA128100050
Price (MYR)
264.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
264.00
EA
Add to cart
Specifications
Chemical Name or Material2-Naphthaldehyde
CAS66-99-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0