Methyl sulfone, 98%
Methyl sulfone, 98%
Methyl sulfone, 98%
Thermo Scientific Chemicals
Methyl sulfone, 98%
CAS: 67-71-0 | C2H6O2S | 94.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA127785000
Price (MYR)
700.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
700.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfone
CAS67-71-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec