1-Methylcyclohexanol, 96%
1-Methylcyclohexanol, 96%
1-Methylcyclohexanol, 96%
1-Methylcyclohexanol, 96%
Thermo Scientific Chemicals
1-Methylcyclohexanol, 96%
CAS: 590-67-0 | C7H14O | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA126601000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Methylcyclohexanol
CAS590-67-0
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0