Methyl acetoacetate, 99+%
Methyl acetoacetate, 99+%
Methyl acetoacetate, 99+%
Methyl acetoacetate, 99+%
Thermo Scientific Chemicals
Methyl acetoacetate, 99+%
CAS: 105-45-3 | C5H8O3 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA126150010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl acetoacetate
CAS105-45-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.