4-Methoxybenzylamine, 98%
4-Methoxybenzylamine, 98%
4-Methoxybenzylamine, 98%
4-Methoxybenzylamine, 98%
Thermo Scientific Chemicals
4-Methoxybenzylamine, 98%
CAS: 2393-23-9 | C8H11NO | 137.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA125780250
Price (MYR)
314.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
314.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Methoxybenzylamine
CAS2393-23-9
Health Hazard 1GHS Signal Word: Danger
ColorColorless to Yellow
Density1.0500g/mL