3'-Methoxyacetophenone, 98%
3'-Methoxyacetophenone, 98%
3'-Methoxyacetophenone, 98%
3'-Methoxyacetophenone, 98%
3'-Methoxyacetophenone, 98%
CAS: 586-37-8 | C9H10O2 | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
Packaging:
Glass bottle
Catalog number FSA125680500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3'-Methoxyacetophenone
Name Note98%
CAS586-37-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning