DL-Methionine, 99+%
DL-Methionine, 99+%
DL-Methionine, 99+%
Thermo Scientific Chemicals
DL-Methionine, 99+%
CAS: 59-51-8 | C5H11NO2S | 149.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
Packaging:
Plastic Bottle
Catalog number FSA125652500
Price (MYR)
383.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
383.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDL-Methionine
Name Note99+%
Ammonium1000ppm max.
CAS59-51-8
pH5.4 to 6.1 (2% w/v aq. soln.)