Mesitylene, 99%, Extra Pure
Mesitylene, 99%, Extra Pure
Mesitylene, 99%, Extra Pure
Mesitylene, 99%, Extra Pure
Thermo Scientific Chemicals
Mesitylene, 99%, Extra Pure
CAS: 108-67-8 | C9H12 | 120.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA125582500
Price (MYR)
455.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
455.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMesitylene
CAS108-67-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.<br
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea