2-Mercaptobenzothiazole, 95%
2-Mercaptobenzothiazole, 95%
2-Mercaptobenzothiazole, 95%
2-Mercaptobenzothiazole, 95%
Thermo Scientific Chemicals
2-Mercaptobenzothiazole, 95%
CAS: 149-30-4 | C7H5NS2 | 167.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Plastic bottle
Catalog number FSA125452500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Mercaptobenzothiazole
CAS149-30-4
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Indicator
RUO – Research Use Only