D-Mannitol, 98+%
D-Mannitol, 98+%
D-Mannitol, 98+%
Thermo Scientific Chemicals
D-Mannitol, 98+%
CAS: 69-65-8 | C6H14O6 | 182.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
2.5 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA125345000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD-Mannitol
CAS69-65-8
Melting Point165.0°C to 170.0°C
ColorWhite
EINECS Number200-711-8