Malononitrile, 99%
Malononitrile, 99%
Malononitrile, 99%
Thermo Scientific Chemicals
Malononitrile, 99%
CAS: 109-77-3 | C3H2N2 | 66.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA125271000
Price (MYR)
255.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
255.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMalononitrile
Name Note99%
CAS109-77-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.