Octadecylbenzene, 98%
Octadecylbenzene, 98%
Octadecylbenzene, 98%
Octadecylbenzene, 98%
Thermo Scientific Chemicals
Octadecylbenzene, 98%
CAS: 4445-07-2 | C24H42 | 330.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 g
10 g
Packaging:
Glass bottle
Catalog number FSA124700500
Price (MYR)
5,176.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
5,176.00
EA
Add to cart
Specifications
Chemical Name or MaterialOctadecylbenzene
CAS4445-07-2
CAS Min %97.5
CAS Max %100.0
Melting Point32°C to 36°C