Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nitric acid, for analysis, ca. 65% solution in water, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Plastic bottle
Catalog number FSA124660010
Price (MYR)
2,351.00
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
2,351.00
Each
Specifications
Chemical Name or MaterialNitric acid
ColorColorless
CAS7697-37-2
Melting Point-42.0°C
Boiling Point122.0°C
View more
For analysis
RUO – Research Use Only