Hydrochloric acid, 0.1 N standard solution
Hydrochloric acid, 0.1 N standard solution
Hydrochloric acid, 0.1 N standard solution
Hydrochloric acid, 0.1 N standard solution
Thermo Scientific Chemicals
Hydrochloric acid, 0.1 N standard solution
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
View all
Plastic bottle
Catalog number FSA124200025
Price (MYR)
619.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
619.00
EA
Add to cart
Chemical Identifiers
CAS7647-01-0, 7732-18-5
Specifications
Chemical Name or MaterialHydrochloric acid
ColorColorless
Name Note0.1 N Standard Solution
CAS7647-01-0
Boiling Point100.0°C
View more
Volumetric standard solution
RUO – Research Use Only