Dicalite™ speed plus, Diatomaceous Filter-aid
Dicalite™ speed plus, Diatomaceous Filter-aid
Thermo Scientific Chemicals
Dicalite™ speed plus, Diatomaceous Filter-aid
CAS: 68855-54-9 | CNa2O3 | 105.988 g/mol
Have Questions?
Catalog NumberQuantity
FSA1233800101 kg
Catalog number FSA123380010
Price (MYR)
258.00
BOT
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
258.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDicalite™ speed plus
Density<26.0 lb./ft3 (wet density)
Physical FormPowder
ColorWhite
Quantity1 kg
View more
Filter aid
RUO – Research Use Only