Sodium thiosulfate pentahydrate, 99+%, extra pure
Sodium thiosulfate pentahydrate, 99+%, extra pure
Sodium thiosulfate pentahydrate, 99+%, extra pure
Sodium thiosulfate pentahydrate, 99+%, extra pure
Thermo Scientific Chemicals
Sodium thiosulfate pentahydrate, 99+%, extra pure
CAS: 10102-17-7 | Na2O3S2·5H2O | 248.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
1 kg
2.5 kg
Packaging:
View all
Fibre drum
Catalog number FSA123370250
Price (MYR)
3,028.00
EA
Add to cart
Quantity:
25 kg
Packaging:
Fibre drum
Request bulk or custom format
Price (MYR)
3,028.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium thiosulfate pentahydrate
Melting Point48.5°C
CAS10102-17-7
ColorColorless to White
pH6 to 8.4 (10% soln. at 20°C)
View more
Reduction reagent
RUO – Research Use Only