Acetic acid, sodium salt trihydrate, 99+%, extra pure
Acetic acid, sodium salt trihydrate, 99+%, extra pure
Acetic acid, sodium salt trihydrate, 99+%, extra pure
Acetic acid, sodium salt trihydrate, 99+%, extra pure
Thermo Scientific Chemicals
Acetic acid, sodium salt trihydrate, 99+%, extra pure
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA123240010
Price (MYR)
353.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
353.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAcetic acid, sodium salt trihydrate
CAS127-09-3
Melting Point58.0°C
CAS Min %99.0
CAS Max %100.0